Anti-Ki-67 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 16A8]

Supplier: Biolegend
652421 652422
10020-548EA 165.5 USD
10020-548 10020-550
Anti-Ki-67 Rat Monoclonal Antibody (PB (Pacific Blue)) [clone: 16A8]
Antibodies
Pacific Blue™ anti-mouse Ki-67 [16A8]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: ICFC, IF; Size: 100 μg

Each lot of this antibody is quality control tested by our Ki-67 staining protocol below. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume or 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

* Pacific Blue™ has a maximum emission of 455 nm when it is excited at 405 nm. Prior to using Pacific Blue™ conjugate for flow cytometric analysis, please verify your flow cytometer's capability of exciting and detecting the fluorochrome.

Type: Primary
Antigen: Ki-67
Clonality: Monoclonal
Clone: 16A8
Conjugation: PB (Pacific Blue)
Epitope:
Host: Rat
Isotype: Rat IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR