Anti-CX3CR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SA011F11]

Supplier: Biolegend
149007 149008
10020-332EA 167.13 USD
10020-332 10020-334
Anti-CX3CR1 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: SA011F11]
Antibodies
APC anti-mouse CX3CR1 [SA011F11]; Isotype: Mouse IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.06 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CX3CR1
Clonality: Monoclonal
Clone: SA011F11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR