Anti-GL7 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: GL7]

Supplier: Biolegend
144603 144604
10757-364EA 121.8 USD
10757-364 10757-366
Anti-GL7 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: GL7]
Antibodies
FITC anti-mouse/human GL7 Antigen (T and B cell Activation Marker) [GL7]; Isotype: Rat IgM, κ; Reactivity: Mouse, Human; Apps: FC; Size: 500 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: GL7
Clonality: Monoclonal
Clone: GL7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: Rat IgM, κ
Reactivity: Mouse;Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR