Anti-CXCL10 Mouse Monoclonal Antibody [clone: J034D6]

Supplier: Biolegend
519501 519502
10761-904EA 114.3 USD
10761-904 10761-906
Anti-CXCL10 Mouse Monoclonal Antibody [clone: J034D6]
Antibodies
Purified anti-human CXCL10 (IP-10) [J034D6]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Rhesus; Apps: ICFC, WB; Size: 500 μg

Each lot of this antibody is quality control tested by intracellular immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 0.5 μg per million cells in 100 μl volume. For Western blotting, the suggested use of this reagent is 0.5 - 2.0 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CXCL10
Clonality: Monoclonal
Clone: J034D6
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG2a, κ
Reactivity: Human;Cross-Reactivity: Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR