Anti-CD324 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: DECMA-1]

Supplier: Biolegend
147307 147308
10756-584EA 137.91 USD
10756-584 10756-586
Anti-CD324 Rat Monoclonal Antibody (Alexa Fluor® 647) [clone: DECMA-1]
Antibodies
Alexa Fluor® 647 anti-mouse/human CD324 (E-Cadherin) [DECMA-1]; Isotype: Rat IgG1, κ; Reactivity: Mouse, Human, Cross-Reactivity: Cynomolgus, Dog, Swine; Apps: FC, IF, IHC-F; Size: 100 μg

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 1.0 μg per million cells in 100 μl volume. For immunofluorescence microscopy, a concentration range of 2-10 μg/ml is recommended. For immunohistochemical staining on frozen tissue sections, a concentration range of 5 - 10 μg/ml is suggested. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD324
Clonality: Monoclonal
Clone: DECMA-1
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rat
Isotype: Rat IgG1, κ
Reactivity: Mouse;Human;Cross-Reactivity: Cynomolgus;Dog;Swine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR