NewBlot™ Stripping Buffers
Supplier: Li-Cor
928-40030
928-40032
928-40028
102673-518EA
206.89
USD
102673-518
102673-520
102971-254
NewBlot™ Stripping Buffers
Buffers
Electrophoresis Buffers
Stripping Buffers
NewBlot Stripping Buffers are optimized for near-infrared (NIR) Western blots, including blots detected with IRDye® secondary antibodies
- Ideal for removal of secondary antibody after detection of highly abundant targets. See performance table for stripping buffer comparisons.
- Optimized for stripping NIR Western blots detected with IRDye 800CW, IRDye 680LT, or IRDye 680RD secondary antibodies.
- NewBlot PVDF Buffer should not be used with IRDye 700DX conjugates.
Stripping and reprobing a Western blot is a common laboratory method. Many labs choose to make their own stripping buffer. What else could you be doing with that time?