Resprep® SPE Cartridges (Bonded Reversed Phases), Restek
Supplier: Restek
24051 26030 26034 28959 28960 28961 28963
RK24051PK 238.78 USD
RK24051 RK24052 10059-486 10059-488 10015-400 10015-402 76457-578 76457-580 76457-582 76457-584
Resprep® SPE Cartridges (Bonded Reversed Phases), Restek
Chromatography Columns
Resprep® SPE cartridges contain hydrophobic (nonpolar) silica-based adsorbents used to extract hydrophobic analytes from polar matrices, such as water (e.g., pesticides from water).
  • High load
  • Endcapped
These cartridges are for use during stationary phase liquid chromatography.