Spirometer Flow Head
Supplier: Vernier Software & Technology
SPR-FLOW
470219-330EA 59.99 USD
470219-330
Spirometer Flow Head
Educational Materials Physics Educational Materials Mechanics Learning Activities
An extra flow head eliminates the need for equipment downtime while sterilizing an original flow head. The flow head consists of a polycarbonate outer body with a stainless-steel inner mesh.