Sodium glucoheptonate dihydrate ≥98.5%
Supplier: Thermo Scientific Chemicals
Synonyms:
Glucosecarboxylic Acid Sodium Salt, Glucoheptonic Acid Sodium Salt
J66811-22
J66811-36
J66811-A1
10815-848EA
47.5
USD
10815-848
10815-850
10815-858
Sodium glucoheptonate dihydrate ≥98.5%
Sodium glucoheptonate dihydrate
CAS No.: 31138-65-5
Molecular Formula: C7H13O8Na
Formula Weight: 248.16
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Solubility: Freely soluble in water
MDL No.: MFCD00013425
Formula:
C₇H₁₄O₈ MW: 248.16 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00013425 CAS Number: 31138-65-5 EINECS: 250-480-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...