Sodium galacturonate anhydrous, anhydrous 98%

Supplier: Thermo Scientific Chemicals

Synonyms: D-(+)-Galacturonic acid sodium salt

J66724-06 J66724-14
10814-798EA 78.3 USD
10814-798 10814-800
Sodium galacturonate anhydrous, anhydrous 98%
Sodium galacturonate anhydrous

Purity: 98% CAS No.: 14984-39-5 Molecular Formula: C6H9NaO7 Formula Weight: 216.12 Physical Form: Powder Sensitivity: Hygroscopic Appearance: White to off-white MDL No.: MFCD06201013

Formula: C₆H₉NaO₇
Storage Temperature: Ambient
MDL Number: MFCD06201013
CAS Number: 14984-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR