Puerarin 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
8-(beta-D-Glucopyranosyl)-4',7-dihydroxyisoflavone
Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
Purity: 98%
CAS No.: 3681-99-0
Molecular Formula: C21H20O9
Formula Weight: 416.38
Physical Form: Powder
Appearance: White to off-white
Melting Point: 187-189°
Solubility: Soluble in DMSO or DMF. Slightly soluble in water or ethanol
Beilstein Reference: 64198
MDL No.: MFCD00076007
Thought to interfere with the synthesis of mycolic acids, an essential fatty acid component found in Mycobacterium cell walls
Formula:
C₂₁H₂₀O₉ MW: 416.38 g/mol Melting Pt: 187…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD00076007 CAS Number: 3681-99-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...