Lactobionic acid 97% (by titrimetric analysis)
Supplier: Thermo Scientific Chemicals
Synonyms:
(2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanoic acid, 4-O-β-D-Galactopyranosyl-D-gluconic acid
J66368-14
J66368-22
J66368-30
10814-584EA
81.2
USD
10814-584
10814-586
10814-588
Lactobionic acid 97% (by titrimetric analysis)
Lactobionic acid
Purity: 97%
CAS No.: 96-82-2
Molecular Formula: C12H22O12
Formula Weight: 358.30
Physical Form: Powder
Appearance: White to off-white
Odor: Odorless
Melting Point: 113-118°
Solubility: Soluble in water
Merck Reference: 14,5342
Optical Rotation: 25°(c=10 in water)
MDL No.: MFCD00078147
Formula:
C₁₂H₂₂O₁₂ MW: 358.30 g/mol Melting Pt: 113…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00078147 CAS Number: 96-82-2 EINECS: 202-538-3 Merck Index: 14,5342 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...