3-Bromo-7-nitro-1H-indazole 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Inhibitor of nitric oxide synthase (NOS)
Purity: 98+%
CAS No.: 74209-34-0
Molecular Formula: C7H4BrN3O2
Formula Weight: 242.00
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Sensitivity: Light Sensitive
Appearance: Pale yellow
Solubility: Soluble in ethanol at 25mg/ml. Also soluble in DMSO
MDL No.: MFCD00159910
Inhibitor of nitric oxide synthase (NOS)
Formula:
C₇H₄BrN₃O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD00159910 CAS Number: 74209-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...