Tellurium, Plasma Standard Solution, Specpure®, Te 10 µg/ml, Thermo Scientific

Supplier: Thermo Scientific
Specpure®
045265-AE
AA45265-AEEA 217.99 USD
AA45265-AE
Tellurium, Plasma Standard Solution, Specpure®, Te 10 µg/ml, Thermo Scientific
Standards Cationic Standards Tellurium Standards
Molecular Formula: Te in 2% HNO3
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134062
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR