Quartz, rods Ø 10.0 mm (0.394 in), fused

Supplier: Thermo Scientific Chemicals
Danger

042294-NK 042294-NT
AA42294-NKEA 32.82 USD
AA42294-NK AA42294-NT
Quartz, rods Ø 10.0 mm (0.394 in), fused
Quartz

CAS No.: 14808-60-7 Physical Form: Rod Appearance: Colorless Odor: Odorless Melting Point: 1710° Boiling Point: 2230° MDL No.: MFCD00217788

Formula: SiO₂
Boiling Pt: 2230 °C
Melting Pt: 1710 °C
Storage Temperature: Ambient
MDL Number: MFCD00217788
CAS Number: 14808-60-7
EINECS: 238-878-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5273273c3b2e473c6ced4e97e18a338f3d21302c&vid=9c414ebb18d830bfa8a77b318dbbb0e6a96da07b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR