Glassy carbon, spherical powder 400-630 µm Type2
Supplier: Thermo Scientific Chemicals
|
Warning
|
041481-30
041481-18
AA41481-30EA
486
USD
AA41481-30
AA41481-18
Glassy carbon, spherical powder 400-630 µm Type2
Glassy carbon
UN No.: 1325
MDL No.: MFCD00133992
Formula:
C MW: 12.01 g/mol Melting Pt: 3652 Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 UN: 1325 ADR: 4.1 III Merck Index: 13,01818 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...