Anti-WIF1 Rat Monoclonal Antibody [clone: 134714]

Supplier: R&D Systems
MAB135-500 MAB135-050
220019-022EA 2876.4 USD
220019-022 220019-019
Anti-WIF1 Rat Monoclonal Antibody [clone: 134714]
Antibodies
Type: Primary
Antigen: WIF1
Clonality: Monoclonal
Clone: 134714
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR