Anti-LILRB2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 287219]

Supplier: R&D Systems
FAB2078A-100 FAB2078A-025
102881-392EA 548.09 USD
102881-392 102881-726
Anti-LILRB2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 287219]
Antibodies
Type: Primary
Antigen: LILRB2
Clonality: Monoclonal
Clone: 287219
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR