ERGOMAT® Infinity Bubble Mats, White, Ergomat
Supplier: Ergomat
IN0203-W IN0204-W IN0206-W IN0209-W IN0212-W IN0303-W IN0304-W IN0306-W IN0309-W IN0312-W IN0404-W IN0406-W IN0409-W IN0412-W
89170-108EA 197.45 USD
89170-108 89170-110 89170-114 89170-120 89170-126 89170-144 89170-146 89170-150 89170-156 89170-162 89170-180 89170-184 89170-190 89170-196
ERGOMAT® Infinity Bubble Mats, White, Ergomat
Floor Mats Traction and Anti-Fatigue Floor Mats
ERGOMAT Infinity Bubble mats deliver the all the ergonomic comfort of polyurethane mats with enhanced durability which results from the addition of a thin, non-porous, closed cell polyurethane membrane on the upper surface of the mat
This silicone and latex free mat is anti-static, UV resistant and certified for use in ISO Class 5 (100) cleanrooms. The white mat also improves lighting in a work area and can reduce energy costs by reflecting rather than absorbing light.
This addition gives the Infinity a degree of chemical and moisture resistance as well as more ready cleanability that is not matched by a typical open cell molded polyurethane mat.
Ordering information: Infinity Bubble mats are available in Black or White and can be made in any shape and size, and every edge will always be beveled for safety. The ERGOMAT Infinity Bubble is recommended for use in a dry, non-abrasive environments and has a 6 year manufacturer’s warranty when used according to manufacturer’s guidelines.