Merit™ Adson-Brown Tissue Forceps, Physician Grade, Sklar
Supplier: Sklar
97-603 97-778
10184-872EA 43 USD
10184-872 10184-908
Merit™ Adson-Brown Tissue Forceps, Physician Grade, Sklar
Forceps
Merit™ Adson-Brown Tissue Forceps are for holding and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow. They are similar to standard Adson forceps, however, Adson-Brown forceps feature multiple rows of teeth, typically 7 x 7 or 9 x 9. They are used to grasp delicate tissue, most commonly in plastic surgery. Merit™ instruments are Sklar’s office-grade line of products and most commonly utilized for routine and in-office minor procedures.
High quality instruments crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices, that require quality, durable instruments.