Pierce™ Detergent Removal Spin Column, Thermo Scientific

Supplier: 0000013509

87776 87777 87778 87779 87780
PI87776EA 292.92 USD
PI87776 PI87777 PI87778 PI87779 PI87780
Pierce™ Detergent Removal Spin Column, Thermo Scientific
Protein Extraction Protein Extraction Reagents

Pierce™ Detergent Removal Resin efficiently binds to and removes high concentrations of detergents from 10 µL to 1 mL samples with minimal sample loss.


  • Fast and efficient protocol
  • Removes greater than 95% of detergents with high protein recovery
  • Helps improves mass spectrometry peptide coverage
  • Best for processing samples with proteins or peptides at greater than 100 µg/mL


These microcentrifuge columns come prepacked with 125 µL of proprietary resin.


Ordering information: Upon receipt store at 4°C. The resin is stored in 0.15 M NaCl, 5% sodium azide.


Caution: For Research Use Only. Not for use in diagnostic procedures.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR