(±)-1,2-Propanediol ≥99.5% ACS

Supplier: Electron Microscopy Sciences
Warning

Synonyms: Propylene glycol, 1,2-Propanediol, (±)-1,2-Propylene glycol, 1,2-Dihydroxypropane, (±)-1,2-Dihydroxy propane

20352 20350
100504-250EA 170.07 USD
100504-250 100504-248
(±)-1,2-Propanediol ≥99.5% ACS
Propylene glycol

(1,2-Propanediol) CH3CHOHCH2OH F.W. 76.10 CAS 57-55-6 Specific Gravity (H2O=1): 1.04 Assay99.5%

Formula: CH₃CH(OH)CH₂OH
MW: 76.1 g/mol
Boiling Pt: 187.6
Melting Pt: -59
Density: 1.0362 g/cm³ (8.647 lbs/gal.)
Flash Pt: >100 °C (>212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064272
CAS Number: 57-55-6
Merck Index: 13,07947
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4c926bc695362012d63f56f50f1085dca6c07056&vid=4c926bc695362012d63f56f50f1085dca6c07056&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.5%
Chloride 1 ppm
Residue after ignition 0.005%
Titratable acid 0.0005 meq/g
Water 0.2%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR