Acetonedimethyl acetal

Supplier: Electron Microscopy Sciences
Danger

Synonyms: 2,2-Dimethoxypropane, Dimethoxypropane, DMP, Dimethoxy-2,2-propane

13281 13280
100503-824EA 244.9 USD
100503-824 100503-822
Acetonedimethyl acetal
Acetone-dimethyl acetal

Used as a dehydration agent.


Miller, L.L. and Jacks, T.J. (1975). Rapid chemical dehydration of samples for electron microscopic examinations. J. Histochem. Cytochem, 23:107
Thorpe J.R. and Harvey D.M.R. (1979). Optimization and investigation of the use of 2,2-dimethoxypropane as a dehydration agent for plant tissue in TEM. J. Ultrastrut. Res. ,68:186
ry, 76:407
Beckmann H.J. and Dierichs, R. (1982). Lipid extracting properties of 2,2-Dimethoxypropane as revealed by electron microscopy and thin layer chromatography Histochemist

Formula: C₅H₁₂O₂
Boiling Pt: 83 °C
Density: 0.85 g/cm³ (7.093 lbs/gal.)
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008479
CAS Number: 77-76-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f47ddd751b5312b78b2c91d96f14ef4a3f32f4c1&vid=f47ddd751b5312b78b2c91d96f14ef4a3f32f4c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity 0.849


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR