Dibutyl phthalate

Supplier: Electron Microscopy Sciences
Danger

Synonyms: DBP, Phthalic acid dibutyl ester

13110 13100 13101
100503-812EA 52.16 USD
100503-812 100503-810 100504-854
Dibutyl phthalate
Dibutyl phthalate

A plasticizer for epoxy resins.

Formula: C₁₆H₂₂O₄
MW: 278.35 g/mol
Boiling Pt: 340 °C (644 °F)
Melting Pt: –35.8 °C (–32 °F)
Density: 1.049 g/cm³
Flash Pt: 188 °C (370 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00009441
CAS Number: 84-74-2
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=66844e1f89391baa6e06b0be7425ebdda8b6624c&vid=66844e1f89391baa6e06b0be7425ebdda8b6624c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific gravity 1.049

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR