Lahey Forceps, OR Grade, Sklar
Supplier: Sklar
17-3255 17-3355
10194-788EA 149.16 USD
10194-788 10194-792
Lahey Forceps, OR Grade, Sklar
Forceps
Sklar's® Lahey Forceps are a hemostats commonly used for clamping blood vessels or tissue before cauterization or ligation. They have ratcheted, finger ring handles. Horizontal serrations the entire length of the jaw, and jaws half the length of the shank make this instrument unique.
Lahey’s with shorter jaws may also be used for soft tissue dissection typically no deeper than 6-8 inches, or in laparotomy, thyroid and vaginohysterectomy procedures.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.