Sklarlite™ Littauer Cutting Forceps, OR Grade, Sklar
Supplier: Sklar
23-9251 23-9255 23-9259 23-9273 23-9275 23-9276
10195-382EA 246.72 USD
10195-382 10195-384 10195-386 10195-388 10195-390 10195-392
Sklarlite™ Littauer Cutting Forceps, OR Grade, Sklar
Bone Cutters and Rongeurs
Sklarlite™ Littauer Cutting Forceps are a lighter version of the standard, non-ratcheted, plier handle instrument for cutting nail and bone
The short, sharp blades are able to produce enough force to cut through thick nail and small bone, but delicate enough not to cause trauma to surrounding tissue or organs. This is helpful with nail debridement which should be completed in short, small sections to avoid damage to the hyponychium.
This lightweight modification provides the user with greater control and less fatigue. As nail nippers they are used for nail debridement. They may also be used to nip delicate bones found in the facial and nasal region, or small animal (mice) veterinary procedures.