Human Recombinant NAP-2 (from E. coli)
Supplier: Peprotech
300-14-2UG 300-14-10UG 300-14-100UG 300-14-250UG 300-14-500UG 300-14-1MG
10780-488EA 132.81 USD
10780-488 10780-490 10780-492 10780-494 10780-496 10780-498
Human Recombinant NAP-2 (from E. coli)
Proteins and Peptides
NAP-2 is a CXC chemokine that can signal through the CXCR1 and CXCR2 receptors. It is produced in leukocytes by enzymatic processing of a precursor called platelet basic protein (PBP). NAP-2 chemoattracts and activates neutrophils. Recombinant Human NAP-2 (CXCL7) protein is a 7.6 kDa protein containing 70 amino acid residues, including the four highly conserved cysteine residues present in CXC chemokines, and also the “ELR” motif, common to CXC chemokines that bind to CXCR1 and CXCR2.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.