Human Recombinant FGF-21 (from E. coli)
Supplier: Peprotech
100-42-5UG 100-42-25UG 100-42-100UG 100-42-250UG 100-42-500UG 100-42-1MG
10778-990EA 139.89 USD
10778-990 10778-992 10778-994 10778-996 10778-998 10779-000
Human Recombinant FGF-21 (from E. coli)
Proteins and Peptides
FGF-21 is a secreted growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-21, in the presence of β-Klotho as a protein cofactor, signals through the FGFR 1c and 4 receptors, and stimulates insulin-independent glucose uptake by adipocytes. Recombinant Human FGF-21 is a 19.5 kDa protein containing 182 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.