Human Recombinant IL-1RA (from E. coli)
Supplier: Peprotech
AF-200-01RA-20UG AF-200-01RA-100UG AF-200-01RA-250UG AF-200-01RA-500UG AF-200-01RA-1MG
10778-230EA 183.06 USD
10778-230 10778-232 10778-234 10778-236 10778-238
Human Recombinant IL-1RA (from E. coli)
Proteins and Peptides
Interleukin-1 receptor antagonist (IL-1RA) is a naturally-occurring, inflammatory-inhibitor protein. It inhibits the activity of IL-1α and IL-1β by competitively blocking their binding to type I and type II receptors. IL-1RA is produced by corneal epithelial cells, monocytes, neutrophils, macrophages, and fibroblasts. Therapeutically, IL-1RA may help in the treatment of sepsis, cachexia, rheumatoid arthritis, chronic myelogenous leukemia, asthma, psoriasis, and inflammatory bowel disease. Recombinant Human IL-1RA is a 17.2 kDa protein consisting of 153 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.