Human Recombinant VEGF (from E. coli)
Supplier: Peprotech
AF-100-20-2UG AF-100-20-10UG AF-100-20-100UG AF-100-20-250UG AF-100-20-500UG AF-100-20-1MG
10778-008EA 181.47 USD
10778-008 10778-010 10778-012 10778-014 10778-016 10778-018
Human Recombinant VEGF (from E. coli)
Proteins and Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through the three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant Human VEGF
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.