Rat Recombinant IL-9 (from E. coli)
Supplier: Peprotech
400-18-2UG 400-18-10UG 400-18-100UG 400-18-250UG 400-18-500UG 400-18-1MG
10774-524EA 132.81 USD
10774-524 10774-526 10774-528 10774-530 10774-532 10774-534
Rat Recombinant IL-9 (from E. coli)
Proteins and Peptides
IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes. IL-9 enhances the proliferation of T lymphocytes, mast cells, erythroid precursor cells and megakaryoblastic leukemia cell lines. Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin’s disease. Recombinant Rat IL-9 is a 14.3 kDa protein of 127 amino acid residues, including 10 cysteine residues that are fully conserved between the human and murine proteins.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.