Human Recombinant MIP-5 (from E. coli)
Supplier: Peprotech
300-43-5UG
300-43-25UG
300-43-50UG
300-43-100UG
300-43-250UG
300-43-500UG
300-43-1MG
10773-966EA
139.89
USD
10773-966
10773-968
10773-970
10773-972
10773-974
10773-976
10773-978
Human Recombinant MIP-5 (from E. coli)
Proteins and Peptides
MIP-5 is a CC chemokine that is expressed in the heart, skeletal muscle and adrenal gland. MIP-5 primarily signals through the CCR1 receptor, but has also been found to bind to CCR3. MIP-5 is chemotactic towards T cells and monocytes. Recombinant Human MIP-5 (CCL15) is a 10.1 kDa protein containing 92 amino acid residues, including the four highly conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.