Human Recombinant MDC (from E. coli)
Supplier: Peprotech
300-36A-5UG 300-36A-20UG 300-36A-100UG 300-36A-250UG 300-36A-500UG 300-36A-1MG
10773-884EA 139.89 USD
10773-884 10773-886 10773-888 10773-890 10773-892 10773-894
Human Recombinant MDC (from E. coli)
Proteins and Peptides
MDC is a CC chemokine that is produced in B cells, macrophages, monocyte-derived dendritic cells, activated NK cells and CD4 T cells. It signals through the CCR4 receptor. MDC chemoattracts monocytes, dendritic cells and NK cells, and exerts HIV-suppressive activity. The 67 amino acid form of MDC displays reduced chemoattractant activity, but retains HIV-suppressive activity. Recombinant Human MDC (CCL22) is an 8.1 kDa protein containing 69 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.