Human Recombinant MCP-3 (from E. coli)
Supplier: Peprotech
300-17-2UG 300-17-10UG 300-17-50UG 300-17-100UG 300-17-250UG 300-17-500UG 300-17-1MG
10773-584EA 132.81 USD
10773-584 10773-586 10773-588 10773-590 10773-592 10773-594 10773-596
Human Recombinant MCP-3 (from E. coli)
Proteins and Peptides
The MCP proteins are members of the CC chemokine family that signal through CCR2 and, with the exception of MCP-1, other CCR receptors. The MCP proteins chemoattract and activate monocytes, activated T cells, basophils, NK cells, and immature dendritic cells. The MCP family cross-reacts across species. Recombinant Human MCP-3 (CCL7) is a 9.0 kDa protein containing 76 amino acid residues including the four highly conserved cysteine residues present in the CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.