Human Recombinant FGF-19 (from E. coli)
Supplier: Peprotech
100-32-5UG 100-32-25UG 100-32-50UG 100-32-100UG 100-32-250UG 100-32-500UG 100-32-1MG
10772-176EA 143.41 USD
10772-176 10772-178 10772-180 10772-182 10772-184 10772-186 10772-188
Human Recombinant FGF-19 (from E. coli)
Proteins and Peptides
The FGF family plays central roles during prenatal development and postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-19, a member of the FGF family, is a high-affinity heparin-dependent ligand for FGFR4. FGF-19 is expressed during brain development and embryogenesis. Recombinant Human FGF-19 is a 21.8 kDa protein containing 195 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.