Nitrile Examination Gloves, Tronex®
Supplier: Tronex
9010-10 9010-20 9010-30 9010-35
10153-084BX 22.84 USD
10153-084 10153-086 10153-088 10153-090
Nitrile Examination Gloves, Tronex®
Gloves
Exclusive nitrile material formulation features a fully textured surface for enhanced durability and abrasion resistance.
  • Fully textured for sustained grip during use
  • Enhanced tensile strength and elasticity with added softness for comfort and reduced hand fatigue
  • Barrier protection from bloodborne pathogens, as well as resistance to chemotherapy drugs