1-Ethynylpyrene 98
Supplier: Thermo Scientific Chemicals
H37327-03
H37327-06
H37327-MD
AAH37327-03EA
120
USD
AAH37327-03
AAH37327-06
AAH37327-MD
1-Ethynylpyrene 98
1-Ethynylpyrene
1-Ethynylpyrene, 98+%
Purity: 98+%
CAS No.: 34993-56-1
Molecular Formula: C18H10
Physical Form: Powder
Appearance: Yellow
Melting Point: 113-116°
Beilstein Reference: 2365008
MDL No.: MFCD02093933
Formula:
C₁₈H₁₀ MW: 226.28 g/mol Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093933 CAS Number: 34993-56-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...