4-Isopropylbenzoyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
H36645-03
H36645-06
AAH36645-03EA
69.66
USD
AAH36645-03
AAH36645-06
4-Isopropylbenzoyl chloride 97%
4-Isopropylbenzoyl chloride
4-Isopropylbenzoyl chloride, 97%
Purity: 97%
CAS No.: 21900-62-9
Molecular Formula: C10H11ClO
Formula Weight: 182.65
Physical Form: Liquid
Sensitivity: Moisture Sensitive
Appearance: Pale yellow
UN No.: 3265
MDL No.: MFCD03424703
Formula:
C₁₀H₁₁ClO MW: 182.647 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03424703 CAS Number: 21900-62-9 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...