Mini 300 Power Supply, G-Biosciences
Supplier: G-Biosciences
BT402
10001-844EA 1102.56 USD
10001-844
Mini 300 Power Supply, G-Biosciences
Power Supplies
In polycarbonate compact and lightweight housing and aluminum base plates, the Mini 300 power supply provides constant voltage or constant current in 1 V or 1mA steps, and it is suited to run both vertical polyacrylamide or horizontal agarose gel electrophoresis experiments.
  • No load detection
  • Shrouded plugs and sockets
Continuous or timed operation (1 to 999 minutes) with alarm function are easily performed using the simple and user-friendly interface, the Mini 300 power supply features two electrode pairs, allowing for two gels to be run simultaneously, saving both time and valuable bench space. With a universal voltage rating, the Mini 300 power supply is also designed and constructed to the most rigorous safety standards.
Certifications: CE, ETL and FCC Marked.