Handguard II® Heavy Duty Knit Cut Resistant Gloves, Wells Lamont
Supplier: Wells Lamont
134852 134853 134854 134855 134856
80090-737EA 44.24 USD
80090-737 80090-739 80090-741 80090-743 80090-745 80090-747
Handguard II® Heavy Duty Knit Cut Resistant Gloves, Wells Lamont
Gloves
Handguard II® heavy duty knit gloves are cut resistant and ambidextrous.
  • Heavy duty knit cut resistant glove
  • Patented combination of high performance fibers, and stainless steel
  • Exceptional protection against cuts and slashes
  • Features Honeywell Spectra® Fiber
  • For use in material handling; meat, poultry and seafood processing
  • Prewashed
  • Preshrunk
Handguard II® heavy duty knit gloves are made from a combination of ultra high strength fibers and stainless steel. They feature Honeywell Spectra® Fiber. These gloves provide protection against cuts and slashes, and they are best for material and food handling applications. Gloves are prewashed and preshrunk for a comfortable fit.
Certifications: ANSI Cut Level 5, A7. CFR 21 – approved for food contact.
Caution: WARNING: Cut resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.