Whizard® ValueSeries Gloves, Wells Lamont
Supplier: Wells Lamont
135027 135028 135029 135030 135031
80076-275EA 25.26 USD
80076-275 80076-277 80076-279 80076-281 80076-283
Whizard® ValueSeries Gloves, Wells Lamont
Gloves
The Whizard ValueSeries gloves are knitted with SpectraGuard™, a high-performance fiber for cut and abrasion resistance.
  • Wireless
  • Strong but Lightweight Spectra Fiber
  • SpectraGuard
The wireless gloves also feature Honeywell Spectra® Fiber
Cut resistant and ambidextrous, the gloves work best in light-duty applications.
Pound for pound, Spectra is 10 times stronger than steel yet lightweight.
Certifications: ANSI Cut Level 4, A3. CFR 21 approved for food contact.
Ordering information: The gloves are sold in multiples of six pieces.
Caution: Caution Information: Cut-resistant gloves are extremely cut resistant but not cut proof. Do not subject to high speed or highly serrated blades. Always disconnect power before cleaning or removing slicer blades.