Anti-CD2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RPA-2.10]

Supplier: Tonbo Biosciences
20-0029-T025 20-0029-T100 20-0029-T500
10753-516EA 122.1 USD
10753-516 10753-518 10753-520
Anti-CD2 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: RPA-2.10]
Antibodies
The RPA-2.10 antibody reacts with human CD2, an approximately 50 kDa glycoprotein, and a member of the Ig superfamily. CD2, also known as LFA-2, is a receptor for CD58 in the human and is expressed on the cell surface of 80-90% of human peripheral blood lymphocytes, a subset of NK cells, and all mature T cells. CD2 mediates lymphocyte adhesion and is involved in T cell activation.

Type: Primary
Antigen: CD2
Clonality: Monoclonal
Clone: RPA-2.10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR