Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 325 mesh Ø 0.036 mm (0.0014 in) Type 316
Supplier: Thermo Scientific Chemicals
Danger
046715-FL 046715-GT 046715-HB
AA46715-FLEA 27.3 USD
AA46715-FL AA46715-GT AA46715-HB
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), gauze woven from wire 325 mesh Ø 0.036 mm (0.0014 in) Type 316
Stainless steel
Wire Cloth
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt% Physical Form: Wire Cloth Odor: Odorless MDL No.: MFCD00148499