2-Morpholino-8-phenylchromone 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
LY294002, LY 294002, 2-Morpholino-8-phenyl-4H-chromen-4-one, 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyran
J67289-LB0
J67289-MB
AAJ67289-LB0EA
88.3
USD
AAJ67289-LB0
AAJ67289-MA
AAJ67289-MB
2-Morpholino-8-phenylchromone 98%
2-Morpholino-8-phenylchromone
Solid
Purity: 98%
CAS No.: 154447-36-6
Molecular Formula: C19H17NO3
Formula Weight: 307.35
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Appearance: White to pale yellow
MDL No.: MFCD00270881
Formula:
C₁₉H₁₇NO₃ MW: 307.35 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00270881 CAS Number: 154447-36-6 |
Specification Test Results
Assay Minimum | 98 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...