Gold 95% supplied in 0.1 mM PBS, nanoparticles 10 nm OD1 (Abs. 520 nm)

Supplier: Thermo Scientific Chemicals

J67029-AC J67029-AE
AAJ67029-ACEA 95.3 USD
AAJ67029-AC AAJ67029-AE
Gold 95% supplied in 0.1 mM PBS, nanoparticles 10 nm OD1 (Abs. 520 nm)
Gold

For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM


Liquid


Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM

Formula: Au
MW: 196.97 g/mol
Boiling Pt: 2800
Melting Pt: 1063
Storage Temperature: Ambient
MDL Number: MFCD00003436
CAS Number: 7440-57-5
Merck Index: 12,04526

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR