Ampicillin 100 mg/ml in aqueous solution ready-to-use

Supplier: Thermo Scientific Chemicals
Danger


Filtered through a 0.2 µm filter.

J66972-AB J66972-AC
AAJ66972-ABEA 182.36 USD
AAJ66972-AB AAJ66972-AC
Ampicillin 100 mg/ml in aqueous solution ready-to-use
Ampicillin

Pale yellow liquid


  • Light sensitive


Storage Temperature: -30°C to -10°C Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Pale yellow Odor: Odorless

Formula: C₁₆H₁₉N₃O₄S
MW: 349.4 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00005175
CAS Number: 69-53-4
EINECS: 200-708-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR