Tiamulin fumarate 95%

Supplier: Thermo Scientific Chemicals
Warning

J66020-MC J66020-ME
AAJ66020-MCEA 60.3 USD
AAJ66020-MC AAJ66020-ME
Tiamulin fumarate 95%
Tiamulin fumarate

Targets Gram-positive bacteria as well as a few mycoplasma species


Powder


Applications: Targets gram positive bacteria as well as a few mycoplasma species

Formula: C₃₂H₅₁NO₈S
Boiling Pt: 563 °C
Melting Pt: 147…148 °C
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00145407
CAS Number: 55297-96-6
EINECS: 259-581-6
Merck Index: 14,9419

Order Now

Specification Test Results

Assay Minimum 95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR