2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Amino-2-chloro-5-(trifluoromethyl)pyridine
H64945-03
H64945-06
AAH64945-03EA
77.7
USD
AAH64945-03
AAH64945-06
AAH64945-MD
2-Chloro-5-(trifluoromethyl)pyridin-3-amine 98%
2-Chloro-5-(trifluoromethyl)pyridin-3-amine
Purity: 98%
CAS No.: 72587-18-9
Molecular Formula: C6H4ClF3N2
Formula Weight: 196.56
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white to brown
Beilstein Reference: 19757170
MDL No.: MFCD07375383
Formula:
C₆H₄ClF₃N₂ Storage Temperature: Ambient |
MDL Number:
MFCD07375383 CAS Number: 72587-18-9 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...