(R)-2-Amino-3-(tritylthio)propanamide 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
H-Cys(Trt)-NH2, H-Cys(Trt)-NH₂, S-Trityl-L-cysteinamide
H63628-03
H63628-06
AAH63628-03EA
46.4
USD
AAH63628-03
AAH63628-06
(R)-2-Amino-3-(tritylthio)propanamide 98%
(R)-2-Amino-3-(tritylthio)propanamide
Purity: 98%
CAS No.: 166737-85-5
Molecular Formula: C22H22N2OS
Formula Weight: 362.49
Physical Form: Powder
Appearance: Off-white
MDL No.: MFCD22126061
Formula:
C₂₂H₂₂N₂OS MW: 362.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22126061 CAS Number: 166737-85-5 |
Specification Test Results
Assay Minimum | 98 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...