Whatman™ Grade GF 6 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Supplier: Cytiva
10370002 10370003 10370005 10370006 10370007 10370008 10370010 10370011 10370012 10370018 10370019 10370020 10370050 10370004
10034-830PK 292.6 USD
10034-830 10034-832 10035-664 10035-666 10035-668 10035-670 10035-672 10035-674 10035-676 10035-678 10035-680 10035-682 10035-684 10035-880
Whatman™ Grade GF 6 Glass Microfiber Filters with Binder, Whatman products (Cytiva)
Filters Glass Fiber Filters
Whatman Grade GF 6 glass microfiber filters from Cytiva's business are suitable for water and wastewater analysis, with extremely fine particle retention and added strength.
  • Sub micron filter retention efficiency to capture extremely small particles
  • Inorganic binder for increased strength
  • Temperature-resistant up to 500 °C
  • Can be used as a pre-filter for dense samples
  • Frequently used in waste water analysis
Grade GF 6 glass filters are suitable for water pollution and waste water analysis. Both require a sub micron filter that can capture tiny particles without weakening. Other applications include: Use as a beer filter in applications including protein removal; Determination of chlorophyll and phytoplankton residue; Determination of filterable substances and residue on ignition; Analysis of aggressive media (e.g., acidic gases); Determination of iron content in the presence of iron oxides; Liquid scintillation measurement.